Learn More
StatLab TISSUE CAPTURE PEN
SDP
Supplier:  StatLab TAT01
Encompass_Preferred
Apply to slides to prevent frozen sections of paraffin sections from falling off. Each pen will coat up to 5000 slides.
Catalog No. NC0321291
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.