Learn More
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-R+(M) (T3741) is used for RI matching while CUBIC-L (T3740) is used for delipidation.
Note: Both T3893 (or T3741) and T3740 are required for the CUBIC clearing process.
Specifications
Specifications
| Capacity | 100 mL |
| For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
| Product Line | CUBIC |
| For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
