Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCI America™ Tissue-Clearing Reagent CUBIC-X1
SDP

Catalog No. T3866100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Capacity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T3866100ML 100 mL
T386625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3866100ML Supplier TCI America™ Supplier No. T3866100ML
Add to Cart
Add to Cart

CUBIC-X1 is an optional addition to the CUBIC clearing process, and can help swell/expand tissues for improved visibility. Used in combination with CUBIC-X2 (T3867).

  • The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
  • CUBIC-X1 is an optional addition to the CUBIC clearing protocol, and can be used to swell/expand tissue for improved visibility.

Note: Both CUBIC-X1 (T3866) and CUBIC-X2 (T3867) are required for the CUBIC clearing process with expansion/swelling.

Specifications

Capacity 100 mL
For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.