Learn More
Med Vet International Miltex Vantage Tissue Forceps, 5.5in, with 1x2 Teeth
Supplier:  Med Vet International MILV9644
Encompass_Preferred
The Miltex Vantage V96-44 Tissue Forceps are made of mid-grade stainless steel and measure 5.5in.They have 1x2 teeth that is used for grasping and securely holding tissue during surgical procedures while minimizing tissue damage.
Catalog No. 50-255-9444
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.