Learn More
Diagnostic Biosystems TISSUE GLUE Tissue Glue
Supplier:  Diagnostic Biosystems K096
Encompass_Preferred
Tissue Glue uses a two-step process to adhere tissues onto a microscope slide. Tissue Glue is particularly useful for maintaining strong attachment of the tissue to the slide during Heat-induced Epitope Retrieval (HIER) in preparation for immunohistochemical staining. The first reagent is applied to the tissue to form attachment points on the slide and on the tissue. The second reagent contains a glue-like substance that covalently bind to the attachment points, thereby firmly attaching the tissue to the slide. Although Tissue-Glue works with all slides, positively charged slides are recommended in order to achieve the optimal results.\n\nThis product is designed for use with both frozen tissue sections and paraffin embedded tissue sections for adhering the tissues firmly onto a glass microscope slide.
Catalog No. NC2049155
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.