Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Tissue Lysis Buffer (TLA) for ReliaPrep™ Large Volume HT gDNA Isolation System

Catalog No. PRA5091 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA5091 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA5091 Supplier Promega Supplier No. A5091
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Lysis buffer composed of Tail Lysis Buffer (TLA) supplied in a 500mL bottle.

  • Used with the ReliaPrep™ Large Volume HT gDNA Isolation System (Cat.# A2751) or the ReliaPrep™ gDNA Tissue Miniprep Systems (Cat.# A2051, and A2052)

Specifications

Content And Storage 15°C to 30°C
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.