Learn More
Fisherbrand™ Tissue Path™ II Cassettes
Catalog No. 15120400L Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue
Lilac
Peach
Pink
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-120-400L Peach
15-120-400D Blue
15-120-400A Yellow
15-120-400E White
15-120-400B Pink
15-120-400G Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15-120-400L
Standard lids fit securely onto cassettes.
  • Molded from a special high-density polymer resistant to all commonly used histological solvents
  • Slanted writing surface accepts markings easily, permitting positive sample identification
Order Info
Packaged in three dispenser boxes of 500
Specifications
Closure Type Separate Lid
Color Peach
Material High Density Polymer
Product Type Tissue Cassette