Description
- Same two-piece molded hinge design as Tissue Path IV Cassettes
- Three-sided writing surface assures positive identification
- Resistant to solvents, decalcifying solutions and compatible with microwave techniques
Order Info
- Packaged in two dispenser boxes, 250 per box
Specifications
Specifications
| Color | Gray |
| Material | Plastic |
| Product Type | Biopsy Cassette |