Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tissue Path™ IV Lids and Bases

Catalog No. 22047067 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Lilac
Peach
Pink
White
Yellow
Product Type:
Embedding Cassette Base
Tissue Cassette Base
Tissue Cassette Lid
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22-047-067 White Tissue Cassette Base
22-047-062 Peach Tissue Cassette Lid
22-047276 Peach Tissue Cassette Base
22-047069 Yellow Tissue Cassette Base
22-047056 White Tissue Cassette Lid
22-047278 Lilac Embedding Cassette Base
22-047060 Pink Tissue Cassette Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-047-067
This item is not returnable. View return policy

For SHUR/Mark™ and other automatic labeling systems

  • Lids are available 1000 per box, two boxes per case
  • Bases are available 500 per box, four boxes per case

Specifications

Color White
Material Acetyl Polymer
Product Type Tissue Cassette Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.