Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Tissue Slicer
SDP

Catalog No. 10000126 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Tissue slicer
Tissue slicer with digital
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-126 Tissue slicer with digital
10-000-127 Tissue slicer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-126 Supplier Stoelting™ Supplier No. 51415
Add to Cart
Add to Cart

Stoelting™ Tissue Slicer is designed to prepare thin slices of living brain and is also excellent for slicing other tissues, blocking fixed tissue for histology, and slicing electrophoretic gels.

Sections as thin as 150 microns can be reliably prepared.

Specifications

For Use With (Application) Neuroscience
Model Tissue slicer with digital
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.