Learn More
Market Lab MasterPress Tissue Tamper, .75X.56 2/PK
Supplier:  Market Lab 56663
Encompass_Preferred
The MasterPress Tissue Tamper by MarketLab is a stainless steel tissue tamper used to flatten and compress tissue samples. It has a flat base plate measuring 3 x 1 inches and a handle that allows the user to apply even pressure. The tamper is autoclavable and can withstand temperatures up to 275°F. It is commonly used in histology labs to prepare tissue samples for microtomy and slide preparation. The tamper allows for quick flattening and evening of tissue samples prior to embedding.
Catalog No. NC1927551
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.