Learn More
Sigma Aldrich TissueFab™ - GelMA-UV bioink
Catalog No. 9054291EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
90-542-91EA 1 Ea.
1 options
Catalog No. 90-542-91EA Supplier Sigma Aldrich Supplier No. 9054291EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Parameters such as pore size, degradation rate, and swell ratio. Temporal and spatial control of the crosslinking reaction can be obtained by adjusting the degree of functionalization and polymerization conditions, allowing for the fabrication of hydrogels with unique patterns, 3D structures, and morphologies. Gelatin methacrylate based bioinks have been used to bioprint osteogenic, chondrogenic, hepatic, adipogenic, vasculogenic, epithelial, endothelial, cardiac valve, skin, tumor and other tissues and constructs.Important tips for optimal bioprinting resultsOptimize printing conditions (e.g., nozzle diameter, printing speed, printing pressure, temperature, cell density) for the features of your 3D printer and your application.Reduce bubble formation. Air bubbles in bioink may hamper bioprinting. Carefully handle the bioink when you mix and transfer it to avoid bubble formation. Do not vortex or shake vigorously.UV light Crosslinking. Position the light source directly above the printed structure. Lower intensity light sources will require shorter distances and longer exposure times to complete crosslinking. Recommended conditions: Place an 800 mW/cm2 light source 8 cm above the printed structure and expose for 30 s.Procedure1. Prepare bioink solution: Warm TissueFab(R) - GelMA-UV bioink in a water bath or incubator set to 37°C for 30 minutes or until the bioink becomes fluid. Gently invert the bioink to make a homogeneous solution. DO NOT vortex or shake vigorously.2. Prepare bioink-cell solution: Resuspend the cell pellet at the desired cell density with the bioink solution by gently pipetting up and down. Typical cell density for extrusion-based bioprinting is 1 to 5 x 106 cells/mL. Load the bioink-cell solution into the desired printer cartridge.3. Bioprint: Cool the filled printing cartridge to 15°C to 19°C to induce gelation, using a temperature controlled printhead or place the cartridge in at 4°C for 10-15 minutes. If print bed temperature control is available, set temperature to 15°C. Follow the 3D printer manufacturer's instructions. Load the print cartridge onto the 3D printer and print directly onto a Petri dish or into multi-well plates. Adjust the flow according to nozzle diameter, printing speed, printing pressure, and temperature.4. Crosslink: Place the UV light source directly above the 3D-bioprinted structure and expose the structure to UV light (wavelength 365 nm). Use the appropriate distance settings and exposure times for your bioprinter. 5. Culture cells: Culture the bioprinted tissue with appropriate cell culture medium following standard tissue culture procedures.TISSUEFAB is a registered trademark of Sigma-Aldrich Co. LLC
Specifications
Quantity 1 Ea.
Synonym Bioink; GelMA; Gelatin methacrylamide; Gelatin methacrylate; Gelatin methacryloyl