Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes in Tube Packs

Catalog No. B851750GN Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851750GN Green
B851750BE Beige
B851750BL Blue
B851750GY Gray
B851750LA Lavender
B851750LG Light Green
B851750LI Lilac
B851750OR Orange
B851750PK Pink
B851750TN Tan
B851750YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. B851750GN Supplier Epredia™ Supplier No. B851750GN
Add to Cart
Add to Cart

For biopsy tissue samples, these cassettes have a back-edge lid hinge, loose-packed lids, and are compatible with all Epredia PrintMate™ Cassette Printers

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Small Squares
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
Color Green
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type TissueLoc Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.