Learn More
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
| Pore Style | Small Squares |
| Quantity | 750/Cs. |
| Closure Included | Yes |
| Closure Type | Separate Lid, Back Hinge |
| PrintMate Compatible | Yes |
| Color | Lilac |
| For Use With (Equipment) | For Use With PrintMate™ Printers |
| Product Type | TissueLoc Biopsy Cassette |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.