Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ TissueLoc™ Cassette IV Slotted Cassettes in Tube Packs

Catalog No. B851720PK Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851720PK Pink
B851720BE Beige
B851720BL Blue
B851720GN Green
B851720GY Gray
B851720LA Lavender
B851720LG Light Green
B851720LI Lilac
B851720OR Orange
B851720TN Tan
B851720WH White
B851720YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. B851720PK Supplier Epredia™ Supplier No. B851720PK
Add to Cart
Add to Cart
This item is not returnable. View return policy

Slotted Cassettes are ideal for use with routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in twelve color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Slots
Quantity 750/Cs.
Closure Included Yes
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
Color Pink
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type TissueLoc™ Slotted Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.