Learn More
Johannes Lieder Tissues and Organs of Domestic Animals, Parasites and Pathogenic Agents
SCI_ED
Catalog No. S02784 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S02784 Cross and Longitudinal Section
1 options
Catalog No. S02784 Supplier Johannes Lieder Supplier No. 7700
Add to Cart
Add to Cart
7700 Tissues and Organs of Domestic Animals, Parasites and Pathogenic Agents, 25 microscope slides
  • 7700 Tissues and Organs of Domestic Animals, Parasites and Pathogenic Agents
  • 25 Microscope Slides
    1. Striated muscle of cow l.s.
    2. Tendon of cow l.s.
    3. Compact bone of cow t.s.
    4. Hyaline cartilage from rib of calf t.s.
    5. Adipose tissue from pig, stained for fat
    6. Liver of pig, t.s.
    7. Small intestine of pig t.s.
    8. Udder (mammary gland) of cow t.s.
    9. Lung of cow t.s.
    10. Bristles of pig w.m.
    11. Skin of pig, l.s. of hair follicles
    12. Tuberculous lung of cow t.s.
    13. Bacillus anthracis, wool sorters disease, smear stained for bacteria
    14. Bacterium erysipelatos, causing red murrain, smear stained for bacteria
    15. Trypanosoma equiperdum, causing dourine in horses, blood smear showing parasites
    16. Eimeria stiedae, coccidiosis, sec. of infected rabbit liver
    17. Dicrocoelium lanceolatum, sheep liver fluke, adult stained and w.m.
    18. Fasciola hepatica, beef liver fluke, ova w.m.
    19. Taenia, tapeworm, proglottids t.s.
    20. Echinococcus granulosus, dog tapeworm, scolices from cyst
    21. Ascaris megalocephala, roundworm of horses, adult female t.s. through midbody
    22. Trichinella spiralis, encysted larvae in muscle l.s.
    23. Sausage t.s.
    24. Paprika, ground
    25. Black pepper, ground
Specifications
Classification Pathogens and Diseases
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Type Microscope Slides
Slide Type Prepared Slide