Learn More
Clontech Labs 3P TITANIUM 1STEP PCR KIT 100RXNS
SDP
Supplier:  Clontech Labs 3P 639504
Encompass
The Titanium One-Step RT-PCR Kit is designed for the reverse transcription of total or poly A+ RNA and the subsequent PCR amplification of a target cDNA in a single reaction tube. The included enzyme mix contains a highly purified Moloney murine leukemia virus (MMLV) reverse transcriptase for first-strand cDNA synthesis, and Titanium Taq DNA Polymerase for PCR amplification. Titanium Taq DNA Polymerase is a blend of a 5'-exonuclease deficient, thermostable DNA polymerase, and TaqStart Antibody for automatic hot start PCR. Reagents for purifying RNA are not supplied.
Catalog No. NC3717685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.