Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium foil, 0.032mm (0.0013in) thick, annealed, 99.7% (metals basis)

Catalog No. AA35881WG
Change view
Click to view available options
Quantity:
23 x 25 cm
23 x 100 cm
23 x 500 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35881WG 23 x 100 cm
AA35881WF 23 x 25 cm
AA35881WH 23 x 500 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35881WG Supplier Thermo Scientific Chemicals Supplier No. 035881WG
Add to Cart

Form: Approx. 8.3g/23 x 25cm | CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Silver to Gray
Melting Point 1660°C
Boiling Point 3287°C
Appearance Silver Gray foil
Density 4.506 g/cm3
Quantity 23 x 100 cm
Odor Odorless
Form Foil
Assay Percent Range 99.7% (Metals basis)
Chemical Name or Material Titanium, Annealed
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.