Learn More
Titanium GE Etch, P4TF8-S12, Class B Alternate, Reagents
Titanium GE Etch, P4TF8-S12, Class B Alternate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$120.00 - $265.00
Chemical Identifiers
| CAS | 7732-18-5 , 7697-37-2 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-335-316
|
Reagents Holdings Llc
CE260200100A |
100 mL |
Each for $120.00
|
|
|||||
|
01-335-315
|
Reagents Holdings Llc
CE2602001A |
1 L |
Each for $265.00
|
|
|||||
|
01-335-314
|
Reagents Holdings Llc
CE260200500A |
500 mL |
Each for $156.00
|
|
|||||
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.