Learn More
Titanium(IV) oxide, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | TiO2 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 79.87g/mol |
| Percent Purity | 99 to 100.5% |
| Grade | puriss. |
| Density | 4.26g/mL at 25°C |
| Chemical Name or Material | Titanium(Iv) Oxide, Meets analytical specification of Ph. Eur.BP, USP |
Safety and Handling
| EINECSNumber | 236-675-5 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.