Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxysulfate Solution 1.9-2.1, For determination of hydrogen peroxyde (H 15), 1.9-2.1%, MilliporeSigma™ Supelco™

Catalog No. 111018783 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8783 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8783 Supplier MilliporeSigma Supelco Supplier No. 342441L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Titanium(IV) oxysulfate is known to form a yellow complex upon reaction with hydrogen peroxide (H2O2) which facilitates the latter′s determination by spectrophotometric methods.

Environmental Food and Beverages and General Analytical
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13825-74-6
Molecular Weight (g/mol) 159.93
MDL Number MFCD00216886

Specifications

Quantity 1 L
Linear Formula TiOSO4
Formula Weight 159.93
Percent Purity 1.9 to 2.1%
Grade Analytical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.