accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHTiHOSOOO

CAS RN 13825-74-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 161.94
Molecular Formula: H2O5STi
InChi Key: ZYPJORZNHDXKSD-UHFFFAOYSA-N
SMILES: O=[Ti].OS(O)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)