Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) tert-butoxide, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s,px6yr1s99s,titanium iv tert-butoxide,titanium tetra-tert-butoxide,titanium tert-butoxide,titanum tetra t-butoxide,tetra-tert-butoxytitanium iv,titanium 4+ 2-methylpropan-2-olate
6451515
2-methylpropan-2-olate;titanium(4+)
CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Ti+4]
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s,px6yr1s99s,titanium iv tert-butoxide,titanium tetra-tert-butoxide,titanium tert-butoxide,titanum tetra t-butoxide,tetra-tert-butoxytitanium iv,titanium 4+ 2-methylpropan-2-olate
6451515
2-methylpropan-2-olate;titanium(4+)
CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Ti+4]
Lewis acid
Flash Point 47°C
Boiling Point 160.0°C to 165.0°C (15.0mmHg)
Packaging Glass bottle
Specific Gravity 0.8893
Quantity 25g
Color Colorless
Density 0.8893g/mL
Assay Percent Range 97.5% min
Linear Formula Ti[OC(CH3)3]4
Beilstein 01, IV, 1613
Formula Weight 340.33
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Titanium(IV) tert-butoxide
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Warning

EINECSNumber : 221-412-9