Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium oxide, 99.9% (metals basis)

Catalog No. AA3900022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3900022 100 g
AA3900014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3900022 Supplier Thermo Scientific Chemicals Supplier No. 03900022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts. | CAS: 12065-65-5 | Ti3O5

Nanocrystalline titanium oxide electrodes is useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12065-65-5
Molecular Formula Ti3O5
MDL Number MFCD00210650
Synonym Trititanium pentoxide

Specifications

Melting Point 1800°C
Physical Form Pieces
Quantity 100 g
Packaging Plastic bottle
Specific Gravity 4.2
Density 4.2
Solubility Information Soluble in hot concentrated sulfuric acid,hydrofluoric acid and alkali.
Percent Purity 99.9%
Chemical Name or Material Titanium oxide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.