Learn More
Eppendorf™ Titermate™ Multichannel Pipetters: Manifolds
Catalog No. 13688170 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
Catalog No. Volume (Metric)
13-688-170 0.5 to 10 μL
1 options
This item is not returnable. View return policy
Catalog No. 13-688-170 Supplier Eppendorf™ Supplier No. 022454005
This item is not returnable. View return policy
Available in 8- and 12-channels
Features a volume adjustment lock that helps prevent user error.
†To add, for example, 12-channel pipetting capability to an 8-channel model, or 8-channel capability to a 12-channel model. Manifolds can also be purchased as replacements.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±8 to 2%
Autoclavable Autoclavable
Number of Channels 8
Volume (Metric) 0.5 to 10 μL
Includes Individual seals and Ceramic pistons