Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TiterTACS In Situ Detection Kit - Colorimetric
GSA_VA

Catalog No. 482296K
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
482296K 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 482296K Supplier R&D Systems™ Supplier No. 482296K
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Proteinase K Solution; Cytonin; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Sapphire; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red

Designed specifically for quantitation of apoptosis in suspension and monolayer cell cultures

Allows quantitative colorimetric analysis of a large number of cell samples using a 96-well plate
Provides quantitation of apoptosis in cultured cells without direct counting of labeled cells
Sensitive and easy to follow assay

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 96 Tests
Content And Storage The product requires storage at both -20 to -70° C and 2 - 8° C. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.