Learn More
BRANDTECH™ BRAND™ Titrating Tube for Titrette™ Bottletop Burette
Catalog No. 13690102 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
10 mL Titrette
Catalog No. For Use With (Equipment)
13-690-102 10 mL Titrette
1 options
Catalog No. 13-690-102 Supplier BRANDTECH™ BRAND™ Supplier No. 707525
Add to Cart
Add to Cart
Includes integrated discharge/recirculation valve.
  • For use with Titrette™ Bottle Top Burette
  • Screw cap fits onto titrating tube to prevent leakage
  • Includes discharge/recirculation valve
TRUSTED_SUSTAINABILITY
Specifications
Product Type Titrating Tube
For Use With (Equipment) 10 mL Titrette
Includes Screw cap for tube, discharge/recirculation tube
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov