Learn More
Medchemexpress LLC TL033 TFA salt | 2746343-70-2 | 98.1% | C76H103N13O23S2 · xC2HF3O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY147340A10MG
Encompass_Preferred
TL033 TFA is a drug-linker conjugate supplied as the trifluoroacetic acid (TFA) salt for antibody-drug conjugate (ADC) research. It pairs a cleavable linker with a bioactive small-molecule toxin for conjugation to antibodies in in vitro and preclinical studies. The material is reported with CAS 2746343-70-2, molecular weight 1630.83 (free base), and supplier-reported purity of ~98.06%.
  • Trifluoroacetic acid salt form suitable for ADC conjugation.
  • Designed as a drug-linker conjugate for antibody-drug conjugate research.
  • High reported purity (≈98.1%).
  • Intended for in vitro and preclinical in vivo conjugation studies.
  • Available in small research pack sizes including 10 mg.
Catalog No. 50-002-58817