Learn More
Beckman Coulter TLA100 ROTOR PACKAGE
Supplier:  Beckman Coulter 343837/Q14360077.0
Encompass
TLA100 rotor package includes:Fixed-Angle, 20 x 0.2 mL rotor, 100,000rpm, 436,000 x g(1) 343775, 200µL, 7mm X 2mm, Thickwall, Polycarbonate (pack of 100)(1) 348302, Tube rack, 7mm(1) 346133, Rotor Vise(1) 927208, Hemostat, 6” curved(1) 824412, O-ring for TLA100 (inner, rotor lid)(1) 824953, O-ring for TLA100 (outer, rotor lid)(1) 306812, Spinkote lubricant(1) 335148, Vacuum grease
Catalog No. NC3919196