Learn More
Atmar Glass Company TLC CAPILLARY SPOTTING TUBES
Supplier:  Atmar Glass Company A1001517
Encompass
TLC, Capillary Spotting Tube, 4 in. L, approx. Pieces Per lb.: 4725, Used for Organic and Water like Solutions, 0.4mm ID x 0.8mm OD for small Spotting
Catalog No. NC9515839
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.