Learn More
Description
Includes
- 1.NB100-56536: TLR6 Antibody (86B1153.2)
- 2.NB100-56563: TLR1 Antibody
- 3.NB100-56566: TLR4 Antibody (76B357.1)
- 4.NB100-56722: TLR2 Antibody (TL2.1)
- 5.NBP1-70343: TLR10 Antibody (3C10C5)
- 6.NBP2-24787: TLR5 Antibody (19D759.2)
Specifications
Specifications
| Antigen | TLR (Cell Surface) Screening |
| Applications | Dot Blot, Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular) |
| Immunogen | TLR1: 25 ug: WB, Flow: H, M, R: Purified TLR2: 25 ug: WB, Flow, IP, IF/ICC: H, D: Purified TLR4: 25 ug: WB, Flow, IHC: H, M, R: Purified TLR5: 25 ug: WB, Flow, IHC: H, M, D: Purified TLR6: 25 ug: Flow, IHC: H: Purified TLR10: 25 ug: Flow: H: Purified WB:Western Blot IHC:Immunohistochemistry IP:Immunoprecipitation IF/ICC:Immunofluorescence/Immunocytochemistry D:Dog H:Human M:Mouse Mk:Monkey R:Rat |
| Quantity | 6 Vials |
| Includes | 1.NB100-56536: TLR6 Antibody (86B1153.2) 2.NB100-56563: TLR1 Antibody 3.NB100-56566: TLR4 Antibody (76B357.1) 4.NB100-56722: TLR2 Antibody (TL2.1) 5.NBP1-70343: TLR10 Antibody (3C10C5) 6.NBP2-24787: TLR5 Antibody (19D759.2) |
| Target Species | Human, Mouse |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
