Learn More
Description
Includes
- 1.NBP2-24729: TLR9 Antibody (26C593.2)
- 2.NBP2-24875: TLR3 Antibody (40C1285.6)
- 3.NBP2-24906: TLR7 Antibody
- 4.NBP2-24917: TLR8 Antibody (44C143)
Specifications
Specifications
| Antigen | TLR (Intracellular) Screening |
| Applications | Dot Blot, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Flow (Intracellular) |
| Immunogen | TLR3: 25 ug: WB, Flow, IP, IHC: H: Purified TLR7: 25 ug: WB, Flow, IHC, IP, IF/ICC: H, M: Purified TLR8: 25 ug: WB, Flow, IHC: H, M: Purified TLR9: 25 ug: WB, Flow, IHC, IF/ICC, CM: H, M, R, RM: Purified WB:Western Blot IHC:Immunohistochemistry IP:Immunoprecipitation CM:Confocal Microscopy FA-Neut Functional Assay Neutralization IF/ICC:Immunofluorescence/Immunocytochemistry D:Dog H:Human M:Mouse Mk:Monkey R:Rat RM:Rhesus Monkey |
| Quantity | 4 Vials |
| Includes | 1.NBP2-24729: TLR9 Antibody (26C593.2) 2.NBP2-24875: TLR3 Antibody (40C1285.6) 3.NBP2-24906: TLR7 Antibody 4.NBP2-24917: TLR8 Antibody (44C143) |
| Target Species | Human, Mouse |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
