Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TLR Screening Antibody Pack
SDP

Catalog No. NBP225083 Shop All Novus Products
Change view
Click to view available options
Quantity:
9 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225083 9 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225083 Supplier R&D Systems™ Supplier No. NBP225083

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Includes

  • 1.NB100-56563: TLR1 Antibody
  • 2.NB100-56720: TLR2 Antibody
  • 3.NB100-56722: TLR2 Antibody (TL2.1)
  • 4.NB100-56723: TLR4 Antibody (HTA125)
  • 5.NB100-56728: TLR2 Antibody (1030A5.138)
  • 6.NBP2-24729: TLR9 Antibody (26C593.2)
  • 7.NBP2-24827: TLR5 Antibody
  • 8.NBP2-24875: TLR3 Antibody (40C1285.6)
  • 9.NBP2-24906: TLR7 Antibody
  • 10.NBP2-24917: TLR8 Antibody (44C143)

A Novus product, part of R&D Systems. For Research applications

TLR Screening specifically detects TLR Screening in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen TLR Screening
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Gene Alias Toll like receptor, Toll-like receptor, Toll-like receptor (TLR)
Immunogen Following are the different antibodies included in the TLR Sampler Kit: TLR1:25 ug:WB, Flow:H, M, R:Purified TLR2 :25 ug:WB, IHC:H, M:Purified TLR2:25 ug:Flow, IP, ICC:H, D:Purified TLR3:25 ug:WB, Flow, IP, IHC:H:Purified TLR4:25 ug:Flow, IP, CM, FA-Neut:H:Purified TLR5:25 ug:WB, Flow, IHC:H, M, R:Purified TLR7:25 ug:WB, Flow, IHC, IP, IF/ICC:H, M:Purified TLR8:25 ug:WB, Flow, IHC:H, M:Purified TLR9:25 ug:WB, Flow, IHC, IF/ICC, CM:H, M, R, RM:Purified WB:Western Blot IHC:Immunohistochemistry IP:Immunoprecipitation CM:Confocal Microscopy FA-Neut Functional Assay Neutralization IF/ICC:Immunofluorescence/Immunocytochemistry D:Dog H:Human M:Mouse Mk:Monkey R:Rat RM:Rhesus Monkey
Quantity 9 Vials
Includes 1.NB100-56563: TLR1 Antibody
2.NB100-56720: TLR2 Antibody
3.NB100-56722: TLR2 Antibody (TL2.1)
4.NB100-56723: TLR4 Antibody (HTA125)
5.NB100-56728: TLR2 Antibody (1030A5.138)
6.NBP2-24729: TLR9 Antibody (26C593.2)
7.NBP2-24827: TLR5 Antibody
8.NBP2-24875: TLR3 Antibody (40C1285.6)
9.NBP2-24906: TLR7 Antibody
10.NBP2-24917: TLR8 Antibody (44C143)
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.