Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody (TL2.1) - BSA Free, R&D Systems™
SDP

Catalog No. NB10056722T
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056722T 0.025 mg
NB10056722 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056722T Supplier R&D Systems Supplier No. NB10056722SS

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody has been used in 61 publication

TLR2 Monoclonal specifically detects TLR2 in Canine, Human, Mouse, Rabbit, Rat samples. It is validated for Block/Neutralize, Cytometric Bead Assay Standard, Dot Blot, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Functional Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro Assay, Western Blot.

Specifications

Antigen TLR2
Applications Blocking Assay, Cytometric Bead Assay Standard, Dot Blot, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Functional Assay, Immunocytochemistry, Immunoprecipitation, In vitro Assay, Western Blot
Classification Monoclonal
Clone TL2.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Flow Cytometry 1 ug/10^6 cells, ELISA reported in multiple pieces of scientific literature, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug, Functional reported in scientific literature (PMID 25412776), In vitro assay reported in scientific literature (PMID 24512642), Dot Blot reported in scientific literature (PMID 27248820), Flow (Cell Surface) reported in scientific literature (PMID 24836676), Block/Neutralize reported in scientific literature (PMID 25412776)
Gene Accession No. O60603
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Gene Symbols TLR2
Host Species Mouse
Immunogen This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7097
Target Species Canine, Mouse, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.