Learn More
Medchemexpress LLC HY-107575 10mg Medchemexpress, TLR4-IN-C34 CAS:40592-88-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10757510MG
Encompass_Preferred
Medchemexpress, HY-107575 10mg TLR4-IN-C34 CAS:40592-88-9 TLR4-IN-C34 is an orally active TLR4 inhibitor and reduces systemic inflammation in models of endotoxemia and necrotizing enterocolitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0715
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.