Learn More
Description
Specifications
Specifications
| Antigen | TLR4 |
| Applications | Flow Cytometry, Immunoprecipitation, Inhibition Assays |
| Classification | Monoclonal |
| Clone | MTS510 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified rat monoclonal antibody IgG2a in PBS without azide. |
| Host Species | Rat |
| Immunogen | Ba/F3 cells expressing mouse TLR4 and MD-2 complex |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.