Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583146 Supplier Invitrogen™ Supplier No. PA583146
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APFEDAGSYS VNLSLAPAGV IRVPAHRPLP GAVPPPLPSA LPAMPSVPTV SSLGGLNFPW MESSRRFVKD RFTA Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX3, also known as homeobox11-like1 (HOX11L2), is a DNA-binding nuclear transcription factor that is expressed in a subset of the primary sensory nervous system. TLX3, along with the related TLX1, is a selector transcription factor that promotes an excitatory glutamatergic neuronal phenotype over an inhibitory GABAergic phenotype opposing LBX1 signals during dorsal spinal cord development. Chromosomal translocations of the TLX3 gene have been shown to result in some forms of T-cell acute lymphoblastic leukemia (T-ALL).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TLX3
Gene Accession No. O43711
Gene Alias homeo box 11-like 2; homeobox 11-like 2; homeobox protein Hox-11L2; Homeobox TLX-3; HOX11L2; Respiratory neuron homeobox protein; RGD1564190; RNX; T cell leukemia, homeobox 3; T-cell leukemia homeobox 3; T-cell leukemia homeobox protein 3; T-cell leukemia, homeobox 3; Tlx1l2; TLX3
Gene Symbols TLX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TLX3. Recombinant protein control fragment (Product #RP-108335).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.