Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TM7SF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7203746 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7660 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7660 Supplier Proteintech Group Inc Supplier No. 120331AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TM7SF2 is involved in the conversion of lanosterol to cholesterol.

Specifications

Antigen TM7SF2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TM7SF2
Gene Accession No. O76062, Q71KT5
Gene Alias Another new gene 1 protein, C 14 sterol reductase, Delta 14 SR, Delta(14) sterol reductase, DHCR14A, NET47, Putative sterol reductase SR 1, Sterol C14 reductase, TM7SF2
Gene Symbols TM7SF2
Host Species Rabbit
Immunogen TM7SF2 Fusion Protein Ag2637
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293688, 7108, 73166
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.