Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TM9SF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569605 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569605 Supplier Invitrogen™ Supplier No. PA569605
  • $587.00
    $489.65 / Each of 1

    Save $97.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

TM9SF3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM9SF3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TM9SF3
Gene Accession No. 0
Gene Alias 1810073M23Rik; 2810031D16Rik; AI115521; AI413748; AW146116; AW549777; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 3; endomembrane protein emp70 precursor isolog; EP70-P-iso; fb75h12; hypothetical protein LOC406698; mKIAA4036; P70-P-iso; RGD1564625; RP11-34E5.1; SM-11044 binding protein; SM-11044-binding protein; SMBP; Tm9sf3; Transmembrane 9 superfamily member 3; Transmembrane 9 superfamily member 3-like protein; transmembrane protein 9 superfamily member 3; UNQ245/PRO282; wu:fa94f05; wu:fb75h12; zgc:56246
Gene Symbols TM9SF3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat TM9SF3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309475
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.