Learn More
Strem, An Ascensus Company CAS# [75-24-1]. 25g. Trimethylaluminum, elec. gr. (99.999+%-Al) PURATREM, 98-1955, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00008252
SDP
Supplier:  Strem, An Ascensus Company 98400825G
Encompass
CAS# [75-24-1]. 25g. Trimethylaluminum, elec. gr. (99.999+%-Al) PURATREM, 98-1955, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00008252. Molecular Weight: 72.09. Molecular Formula: (CH3)3Al. Color/form: colorless liq. Strem# 98-4008. www.strem.com/catalog/v3/98-4008/
Catalog No. 50-197-9366
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.