Learn More
Enzo Life Sciences Trimethylammonio-PTIO. methylsulfate (50mg)
Supplier: Enzo Life Sciences ALX430085M050
Water soluble nitric oxide (NO) spin trap that is non-cell permeable, allowing study of extracellular NO release. Alternative name: TMA-PTIO . CH3SO4-, 3,3,4,4-Tetramethyl-2-trimethylammoniophenyl-2-imidazoline-3-oxide-1-yloxy methylsulfate. Purity: ≥80% (Assay). Appearance: Dark blue crystalline solid. Solubility: Soluble in water, 100% ethanol or DMSO. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.