Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMA16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570925 Supplier Invitrogen™ Supplier No. PA570925
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 91%; Rabbit: 85%; Rat: 100%.

TMA16 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMA16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMA16
Gene Accession No. Q9CR02
Gene Alias 1810029B16Rik; C4orf43; RGD1305222; TMA16; translation machinery associated 16 homolog; translation machinery associated 16 homolog (S. cerevisiae); translation machinery-associated protein 16; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Gene Symbols TMA16
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse TMA16.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66282
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.