Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMB ELISA Buffer Kit, PeproTech®

Catalog No. 900T00
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
900-T00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 900-T00 Supplier Invitrogen™ Supplier No. 900T00
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

This kit contains the key components required for the analysis of 10 Sandwich ELISA plates. It has been specifically formulated to be used in conjunction with PeproTech’s Standard TMB ELISA Development Kits.

  • Contains the key components required for the analysis of ten sandwich ELISA plates
  • Specifically formulated to be used in conjunction with PeproTech standard TMB ELISA development kits.

Included solutions and materials:

  • PBS
  • Wash buffer
  • Blocking buffer
  • Diluent
  • TMB liquid substrate
  • ELISA microplates
  • Sealing film
  • Stop solution
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Buffer
Quantity 1 Kit
Detection Method Colorimetric
For Use With (Application) ELISA
Product Line PeproTech
Substrate Type TMB, HRP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.