Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam TMB ELISA Substrate (Highest Sensitivity), 100 mL

Catalog No. 502582491
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2491 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2491 Supplier Abcam Supplier No. AB171522100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abcam TMB ELISA Substrate (Highest Sensitivity).

  • Species Reactivity: Species independent
  • Storage: Store in the dark

Specifications

Product Type ELISA Substrate
Quantity 100 mL
For Use With (Application) Immunohistochemistry (IHC), ELISA
Content And Storage Store in the dark
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.