Learn More
KEMENTEC SOLUTIONS INC TMB ONE (& prestained) ELISA HRP Substrate, 1L
Supplier:  KEMENTEC SOLUTIONS INC 4380A
Encompass
TMB (3, 3', 5, 5'-tetramethylbenzidine) is a chromogenic substrate for Horseradish Peroxidase (HRP). TMB produces a deep blue color during the enzymatic degradation of hydrogen peroxide by HRP, and the addition of a stop solution gives a clear yellow color that absorbs at 450 nm. This substrate gives higher signal and has a higher sensitivity in ELISA than OPD (ortho-phenylenediamine dichloride). “The Gold Standard” used across the industry. Incredibly stable and robust. Aqueous based – no organic solvents (REACH Annex XVII compliant)
Catalog No. NC2036412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.