Learn More
Medchemexpress LLC HY-11097 5mg Medchemexpress, TMC353121 CAS:857066-90-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY110975MG
Encompass_Preferred
Medchemexpress, HY-11097 5mg TMC353121 CAS:857066-90-1 TMC353121 is a potent respiratory syncytial virus (RSV) fusion inhibitor with pEC50 of 9.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8809
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.