Learn More
Description
Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS Silylation Reagent. It is an excellent adjunct for forming TMS ethers for GC determinations. TMCS is also useful for preparing TMS derivatives of organic acids.
- Excellent adjunct for forming trimethylsilyl ethers for GC determinations
- Used to prepare TMS derivatives of organic acids
Specifications
Specifications
| For Use With | GC |
| Hazards | Flammable, Toxic |
| Storage | Room Temperature |
| Quantity | 25 g |
| Standard Components | Chlorotrimethylsilane |
| Standard Concentration | 98 to 100% |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.