Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675602
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-602 Supplier Invitrogen™ Supplier No. PA141256
  • $587.00
    $487.70 / Each of 1

    Save $99.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: HL-60 cell lysate. Predicted to cross react with mouse and rat. Immunogen sequence: CNYRTQMWDK QKEV.

Involved in vesicular protein trafficking, mainly in the early secretory pathway. targeting. Involved in the maintenance of the Golgi apparatus. Appears to play a role in the biosynthesis of secreted cargo including processing. Involved in endoplasmic reticulum stress response. May play a role in the regulation of heat-shock response and apoptosis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene TMED4
Gene Accession No. Q7Z7H5
Gene Alias 1110014L17Rik; AI326346; endoplasmic reticulum stress-response protein 25; endoplasmic reticulum stress-response protein 25 kDa; ERS25; GMP25iso; HNLF; p24 family protein alpha-3; p24a3; p24alpha3; p26; Putative NF-kappa-B-activating protein 156; putative NFkB activating protein HNLF; RGD1306319; Tmed4; Transmembrane emp24 domain-containing protein 4; transmembrane emp24 protein transport domain containing 4; transmembrane p24 trafficking protein 4
Gene Symbols TMED4
Host Species Rabbit
Immunogen A synthetic peptide containing amino acids (amino acids 55-67) of human HNLF.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.