Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMED5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531580 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531580 Supplier Invitrogen™ Supplier No. PA531580
  • $625.50
    $489.65 / Each of 1

    Save $135.85

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Human (99%), Mouse (91%), Rat (92%), Pig (97%), Chicken (87%), Rhesus Monkey (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The TMED5 gene encodes the transmembrane emp24 domain-containing protein 5, a member of the TMED family, which is involved in intracellular protein trafficking. This family is essential for the transport of proteins between the endoplasmic reticulum and the Golgi apparatus. TMED5 is a member of the γ subfamily and has been shown to regulate vital cellular processes, including protein secretion and signal transduction. Dysregulation of TMED5 has been implicated in various diseases, including hepatocellular carcinoma (HCC). Increased expression of TMED5 mRNA in HCC tissues is associated with poorer overall survival rates compared to normal tissues, suggesting its role in tumor progression and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMED5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene TMED5
Gene Accession No. Q9Y3A6
Gene Alias CGI-100; p24 family protein gamma-2; p24g2; p24gamma2; p28; TMED5; Transmembrane emp24 domain-containing protein 5; transmembrane emp24 protein transport domain containing 5; transmembrane p24 trafficking protein 5; UNQ397/PRO733
Gene Symbols TMED5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TMED5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50999
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.