Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TMEM101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102016 Supplier Invitrogen™ Supplier No. PA5102016
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TMEM101.

May activate NF-kappa-B signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM101
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TMEM101
Gene Accession No. Q91VP7, Q96IK0
Gene Alias 2610511E22Rik; Putative NF-kappa-B-activating protein 130; Tmem101; Transmembrane protein 101
Gene Symbols TMEM101
Host Species Rabbit
Immunogen A synthesized peptide derived from human TMEM101(Accession Q96IK0), corresponding to amino acid residues A115-D165.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76547, 84336
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.